Anti-LYC Rabbit Polyclonal Antibody
Catalog # AGRIAS132709
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:LYC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:His-tagged recombinant part of Arabidopsis thaliana lycopene beta-cyclase, encompassing about 2/3 of the coding region of, AEE74875.1,TAIR: AT3G10230
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: LYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human