Anti-C1-inh Sheep Polyclonal Antibody
Catalog # AGRIAS101314
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:C1INH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Reactivity:Oryza sativa,Spinacia oleracea,Brassica oleracea,Zea mays,A. thaliana,N. tabacum,Solanum lycopersicum,Vitis vinifera,H. vulgare,N. bentamina
- Cross adsorption:No
- Antigen synonyms:C1-INH
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:human C1 Inhibitor CAA30314
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: C1INH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: A. thaliana, Brassica oleracea, H. vulgare, N. bentamina, N. tabacum, Oryza sativa, Solanum lycopersicum, Spinacia oleracea, Zea mays, Vitis vinifera