Anti-Pht1-6 Rabbit Polyclonal Antibody
Catalog # AGRIAS08320
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PHT1-6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:two synthetic peptides from the loop region of Pht1-6 (Q8H6D9), conjugated to KLH
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PHT1-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana