Anti-ZNF341 Rabbit Polyclonal Antibody
Catalog # ORIGTA329642
Leverancier: OriGene
Specificaties
- Antibody type:Primary
- Antigen symbol:ZNF341
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Lyophilized powder. Add 100 ul of distilled water. Final anti-ZNF341 antibody concentration is 1 mg/ml in PBS buffer with 2% sucrose. For longer periods of storage, store at -20C. Avoid repeat freeze-thaw cycles.
- Molecular weight:92 kDa
- Sequence:EGMDNQTVLAVQSLLDGQGAVPDPTGQSVNAPPAIQPLDDEDVFLCGKCK
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:The immunogen for anti-ZNF341 antibody: synthetic peptide directed towards the N terminal of human ZNF341. Synthetic peptide located within the following region: EGMDNQTVLAVQSLLDGQGAVPDPTGQSVNAPPAIQPLDDEDVFLCGKCK
- Size:100 μg
- Pk:100 µG
Specificaties
Over dit artikel
Type: Primary
Antigen: ZNF341
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human