Anti-CALD1 Rabbit Monoclonal Antibody [clone: E-89]
Catalog # ORIGTA327612
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CALD1
- Clonality:Monoclonal
- Clone:E-89
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:The antibody is diluted in tris buffered saline, pH 7,3-7.7, with 1% BSA and <1% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Pk:1 mL
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:25-1:100
Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: E-89
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human