Anti-Shank1 Mouse Monoclonal Antibody [clone: S22-21]
Catalog # ORIGTA326582
Leverancier: OriGene
Specificaties
- Antibody type:Primary
- Antigen name:SH3 and multiple ankyrin repeat domains 1
- Antigen symbol:SHANK1
- Clonality:Monoclonal
- Clone:S22-21
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:469 to 691
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 469-691 (SH3/PDZ domains) of rat Shank1
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specificaties
Over dit artikel
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: Shank1
Clonality: Monoclonal
Clone: S22-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat.