Anti-ATAD1 Mouse Monoclonal Antibody [clone: S125-10]
ORIGTA326468
: OriGene
- Antibody type:Primary
- Antigen name:ATPase family, AAA domain containing 1
- Antigen symbol:ATAD1
- Clonality:Monoclonal
- Clone:S125-10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1 to 361
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 1-361 (Full length) of mouse Thorase,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: ATAD1
Clonality: Monoclonal
Clone: S125-10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat