Anti-EIF4G1 Rabbit Polyclonal Antibody
Catalogus # ORIGTA322698
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:EIF4G1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Ser1232
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:220 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:PepT-ide sequence around phosphorylation site of serine 1232 (pV-S(p)-pL) derived from Human eIF4G.
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: EIF4G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human