Order Entry
Netherlands
ContactUsLinkComponent
Anti-IFNGR2 Rat Polyclonal Antibody [clone: AN-18]
Anti-IFNGR2 Rat Polyclonal Antibody [clone: AN-18]
Catalog # ORIGTA320355
Supplier:  OriGene
Anti-IFNGR2 Rat Polyclonal Antibody [clone: AN-18]
Catalog # ORIGTA320355
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon gamma receptor 2
  • Antigen symbol:
    IFNGR2
  • Clonality:
    Polyclonal
  • Clone:
    AN-18
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Size:
    50 μg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone: AN-18
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse