Order Entry
Netherlands
ContactUsLinkComponent
Colorimetric test kit, chloride, TESTOVAL®
Colorimetric test kit, chloride, TESTOVAL®
  HEYL410527
 :  
Colorimetric test kit, chloride, TESTOVAL®
  HEYL410527
 :  
  • Pk:
    1
  • Description:
    Chloride, colour comparison kit
  • Seq:
    01
  • Range:
    1-5-10-25-50-100 mg/l
Visual colour test method using a comparator disc.