- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG F(c)
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:F(c)
- Cross adsorption:Yes
- Format:Purified IgG
- Form:Clear, colorless liquid
- Antibody fragment:F(ab')2
- Storage buffer:30 mM Triethanolamine, pH 7,2, 5 mM Magnesium Chloride, 0,1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2…8 °C
- Concentration:1,50 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2…8 °C
- Immunogen:Human IgG, Fc fragment
- Purification:Affinity purified using solid phaseHuman IgG
- Size:1,0 mg
- Pk:1 mg
ImmunoReagents Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
- Origin: US
ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human