Anti-AOC3 Rabbit Polyclonal Antibody
Catalog # USBIV2095-03A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:amine oxidase, copper containing 3
- Antigen symbol:AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: AOC3 (amine oxidase, copper containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human