Anti-TRPC4 Mouse Monoclonal Antibody [clone: 10F774]
Catalog # USBIT8666-10B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily C, member 4
- Antigen symbol:TRPC4
- Clonality:Monoclonal
- Clone:10F774
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Rabbit,Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-10 ?g/ml; Immunochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: TRPC4 (transient receptor potential cation channel, subfamily C, member 4)
Clonality: monoclonal
Clone: 10F774
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rabbit