Anti-TEK Rabbit Polyclonal Antibody
USBIT5498-71H
- Antibody type:Primary
- Antigen name:TEK tyrosine kinase, endothelial
- Antigen symbol:TEK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:10000-1:100000
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human