Order Entry
Netherlands
ContactUsLinkComponent
Anti-TCR, V beta 8.1, 8.2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F510]
Anti-TCR, V beta 8.1, 8.2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F510]
Catalog # USBIT2040-25C
Supplier:  US Biological
Anti-TCR, V beta 8.1, 8.2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F510]
Catalog # USBIT2040-25C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TCR V beta 8.1 8.2
  • Clonality:
    Monoclonal
  • Clone:
    10F510
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,5% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: TCR, V beta 8.1, 8.2
Clonality: monoclonal
Clone: 10F510
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse