Anti-TCR, V beta 8.1, 8.2 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F510]
USBIT2040-25B
- Antibody type:Primary
- Antigen symbol:TCR V beta 8.1 8.2
- Clonality:Monoclonal
- Clone:10F510
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,5% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TCR, V beta 8.1, 8.2
Clonality: monoclonal
Clone: 10F510
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse