Anti-Syntaxin 13 Mouse Monoclonal Antibody [clone: 4E212]
USBIS9205-45-100
- Antibody type:Primary
- Antigen name:Syntaxin 13
- Antigen symbol:STX13
- Clonality:Monoclonal
- Clone:4E212
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Hamster,Dog,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Syntaxin 13
Clonality: Monoclonal
Clone: 4E+212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Mouse, Rat