Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]
Catalog # USBIS8000-01B-100
Leverancier: US Biological
Specificaties
- Antibody type:Primary
- Antigen symbol:Substance P
- Clonality:Monoclonal
- Clone:SP-DE4-21
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Guinea pig,Rat,Mouse
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 0,1M glycine.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specificaties
Over dit artikel
Recommended dilution: ELISA: 1:10000; Immunochemistry: 1:300-1:1000
Type: Primary
Antigen: Substance P
Clonality: Monoclonal
Clone: SP-DE4-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig, Human, Mouse, Rat