Anti-Streptococcal Pyrogenic Exotoxin A Rabbit Polyclonal Antibody
Catalog # USBIS7974-06
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Streptococcal Pyrogenic Exotoxin A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Form:lyophilized powder
- Storage buffer:0,01M phosphate, 0,9% sodium chloride, pH 7,4
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Streptococcal Pyrogenic Exotoxin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: