Anti-Semaphorin 3B Rabbit Polyclonal Antibody
Catalog # USBIS0668-48
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 3B
- Antigen symbol:SEMA3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris-glycine, 150mM sodium chloride, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: Semaphorin 3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Zebrafish