Anti-QRFP Pyroglutamylated RFamide Peptide 43 Rabbit Polyclonal Antibody
USBIQ0575-50A
- Antibody type:Primary
- Antigen symbol:QRFP Pyroglutamylated RFamide Peptide 43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:154 to 196
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: QRFP Pyroglutamylated RFamide Peptide 43
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human