Anti-Pyruvate Kinase, Tumor Type M2 Rabbit Polyclonal Antibody
USBIP9545-31
- Antibody type:Primary
- Antigen name:Pyruvate Kinase, Tumor Type M2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
- Recommended dilution: Western Blot: 1:1000 Immunochemistry: 1:100
Type: Primary
Antigen: Pyruvate Kinase, Tumor Type M2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat