Anti-Pyruvate Dehydrogenase Kinase Isoform 4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIP9544-75-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase Kinase Isoform 4
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: Pyruvate Dehydrogenase Kinase Isoform 4
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human