Bestelling invoeren
Netherlands
ContactUsLinkComponent
Anti-Pyruvate Dehydrogenase Kinase Isoform 4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-Pyruvate Dehydrogenase Kinase Isoform 4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBIP9544-75-APC
undefined
Anti-Pyruvate Dehydrogenase Kinase Isoform 4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBIP9544-75-APC

 

  • Type antilichaam:
    Primair
  • Clonality:
    Polyklonaal
  • Conjugatie:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Formulier:
    liquid
  • Opslagbuffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Opslagtemperatuur:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Maat:
    200 µl
  • VPE:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: Pyruvate Dehydrogenase Kinase Isoform 4
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human