Anti-Pyruvate Dehydrogenase E1-alpha Subunit Rabbit Polyclonal Antibody
USBIP9544-15
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase E1-alpha Subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser293
- Storage buffer:Tris-citrate/phosphate, pH 7,5, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Recommended dilution: Western Blot: 1:1000-1:5000
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha Subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human