Anti-POU Domain, Class 2, Transcription Factor 1 Mouse Monoclonal Antibody [clone: 0.T.100]
Catalog # USBIP5250-01D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:POU Domain, Class 2, Transcription Factor 1
- Clonality:Monoclonal
- Clone:0.T.100
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide before the addition of glycerol to 40%.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:250 -1:1000
Type: Primary
Antigen: POU Domain, Class 2, Transcription Factor 1
Clonality: monoclonal
Clone: 0.T.100
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human