Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: P-TYR-01]
USBIP4075-28G
- Antibody type:Primary
- Antigen name:Phosphotyrosine
- Antigen symbol:pY
- Clonality:Monoclonal
- Clone:P-TYR-01
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: P-TYR-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: