Anti-Peroxidase Anti-Peroxidase Rat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIP3350-35
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Peroxidase Anti-Peroxidase
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µl
- Pk:250 µl
Specifications
About this item
Type: Primary
Antigen: Peroxidase Anti-Peroxidase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype:
Reactivity: