Anti-CDKN1B Rabbit polyclonal antibody
USBIP1001-09
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 1B
- Antigen symbol:CDKN1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser10
- Storage buffer:PBS, pH 7,4, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1-3?g/ml; Immunoprecipitation: 5-10 ?g/Immunoprecipitation: reaction; Immunofluorescence: 5-10 ?g/ml
Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat