Anti-Neuron Specific Enolase Mouse Monoclonal Antibody [clone: MIG-N3]
USBIN2175-01F
- Antibody type:Primary
- Antigen name:Neuron Specific Enolase
- Antigen symbol:NSE
- Clonality:Monoclonal
- Clone:MIG-N3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Guinea pig
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSa, 0,1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:500 µl
- Pk:500 µl
Recommended dilution: Immunochemistry: 1:100
Type: Primary
Antigen: Neuron Specific Enolase
Clonality: monoclonal
Clone: MIG-N3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human