Anti-Neurofilament 70, 200kD Mouse Monoclonal Antibody [clone: 5G17]
Catalog # USBIN2160-05
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament 70, 200kD
- Clonality:Monoclonal
- Clone:5G17
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 1% BSA and 0,1% sodium azide.
- Molecular weight:200 kDa
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:5000
Type: Primary
Antigen: Neurofilament 70, 200kD
Clonality: monoclonal
Clone: 5G17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human