Order Entry
Netherlands
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide, 1-10 Rabbit polyclonal antibody
Anti-Brain Natriuretic Peptide, 1-10 Rabbit polyclonal antibody
Catalog # USBIN0520-25C
Supplier:  US Biological
Anti-Brain Natriuretic Peptide, 1-10 Rabbit polyclonal antibody
Catalog # USBIN0520-25C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide, 1-10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Radio Immunoassay: 1:500

Type: Primary
Antigen: Brain Natriuretic Peptide, 1-10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human