Order Entry
Netherlands
ContactUsLinkComponent
Anti-MHC Class 2, DR Mouse Monoclonal Antibody [clone: 10F111]
Anti-MHC Class 2, DR Mouse Monoclonal Antibody [clone: 10F111]
Catalog # USBIM3887-46
Leverancier:  US Biological
Anti-MHC Class 2, DR Mouse Monoclonal Antibody [clone: 10F111]
Catalog # USBIM3887-46
Leverancier:  US Biological

Specificaties

  • Antibody type:
    Primary
  • Antigen name:
    MHC Class 2, DR
  • Clonality:
    Monoclonal
  • Clone:
    10F111
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Sheep,
    Bovine
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    0,2M Tris/HCl, pH 7,4, 5-10% fetal calf serum, 0,01% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    2 ml
  • Pk:
    2 mL

Specificaties

Over dit artikel

Type: Primary
Antigen: MHC Class 2, DR
Clonality: monoclonal
Clone: 10F111
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Sheep