Order Entry
Netherlands
ContactUsLinkComponent
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F104]
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F104]
Catalog # USBIM3887-43D
Supplier:  US Biological
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F104]
Catalog # USBIM3887-43D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MHC Class 2, DQ, DR, Polymorphic
  • Clonality:
    Monoclonal
  • Clone:
    10F104
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Sheep,
    Goat,
    Human,
    Bovine
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:10;

Type: Primary
Antigen: MHC Class 2, DQ, DR, Polymorphic
Clonality: monoclonal
Clone: 10F104
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Goat, Human, Sheep