Order Entry
Netherlands
ContactUsLinkComponent
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F103]
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F103]
Catalog # USBIM3887-43B
Supplier:  US Biological
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F103]
Catalog # USBIM3887-43B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MHC Class 2, DQ, DR, Polymorphic
  • Clonality:
    Monoclonal
  • Clone:
    10F103
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Sheep
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Recommended dilution: Flow Cytometry:1:10;
Do not freeze!

Type: Primary
Antigen: MHC Class 2, DQ, DR, Polymorphic
Clonality: monoclonal
Clone: 10F103
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Sheep