Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-Matrix Metalloproteinase 13 Sheep Polyclonal Antibody
  USBIM2428-09B
Anti-Matrix Metalloproteinase 13 Sheep Polyclonal Antibody
  USBIM2428-09B

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 13
  • Antigen symbol:
    MMP13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    glycine buffered saline, 0,09% sodium azide, 0,1% EACA, 0,01% benzamidine, 1mM EDTA.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Matrix Metalloproteinase 13
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse