Order Entry
Netherlands
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Catalog # USBIM2360-02F-200
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Catalog # USBIM2360-02F-200
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Drosophila,
    Sheep,
    Chicken,
    Human,
    Bovine,
    Xenopus laevis,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000; Flow Cytometry: 10 ?g/ml; Immunochemistry: 1:50

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Drosophila, Human, Mouse, Rat, Sheep, Xenopus laevis