Order Entry
Netherlands
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody (Alexa Fluor® 647/DyLight® 649)
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody (Alexa Fluor® 647/DyLight® 649)
Catalog # USBIM2352-20H
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody (Alexa Fluor® 647/DyLight® 649)
Catalog # USBIM2352-20H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647/DyLight® 649
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Drosophila,
    Sheep,
    Chicken,
    Human,
    Bovine,
    Xenopus laevis,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Flow Cytometry: 10 ?g/ml; Western Blot: 1:5000; Immunochemistry:1:50

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: polyclonal
Clone:
Conjugation: Alexa Fluor® 647/DyLight® 649
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Drosophila, Human, Mouse, Rat, Sheep, Xenopus laevis