Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody (Alexa Fluor® 647) [clone: 10E37]
Catalog # USBIM2352-10B
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody (Alexa Fluor® 647) [clone: 10E37]
Catalog # USBIM2352-10B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    10E37
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Hamster,
    Pig,
    Monkey,
    C. elegans,
    Human,
    Bovine,
    Rat,
    Mouse
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    PBS(pH 7,2), less than 0,1% sodium azide,2mg/ml BSA.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: monoclonal
Clone: 1E+38
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, C. elegans, Hamster, Human, Monkey, Mouse, Pig, Rat, Zebrafish