Order Entry
Netherlands
ContactUsLinkComponent
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M3/84]
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M3/84]
  USBIL9190-12J
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M3/84]
  USBIL9190-12J

 

  • Antibody type:
    Primary
  • Antigen name:
    Lysosome Associated Membrane Protein 2
  • Antigen symbol:
    LAMP2
  • Clonality:
    Monoclonal
  • Clone:
    M3/84
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

 

 

Do not freeze!

Type: Primary
Antigen: Lysosome Associated Membrane Protein 2
Clonality: monoclonal
Clone: M3/84
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse