Order Entry
Netherlands
ContactUsLinkComponent
Anti-LRP, MVP Mouse Monoclonal Antibody [clone: MVP-37]
Anti-LRP, MVP Mouse Monoclonal Antibody [clone: MVP-37]
Catalog # USBIL4500-03J
Supplier:  US Biological
Anti-LRP, MVP Mouse Monoclonal Antibody [clone: MVP-37]
Catalog # USBIL4500-03J
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LRP
  • Clonality:
    Monoclonal
  • Clone:
    MVP-37
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Antigen synonyms:
    MVP
  • Storage buffer:
    0,7%BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    250 ug immunoglobulin/ml
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:20-50; Western Blot: 1:50

Type: Primary
Antigen: LRP, MVP
Clonality: monoclonal
Clone: MVP-37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human