Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # USBIK0199-08J
Supplier:  US Biological
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # USBIK0199-08J
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 8
  • Antigen symbol:
    KRT8
  • Clonality:
    Monoclonal
  • Clone:
    C-51
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Sheep,
    Human,
    Bovine
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: C-51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig, Sheep