Anti-Interleukin 1 Receptor Type II Mouse Monoclonal Antibody [clone: 8.5]
Catalog # USBII7663-25K7
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 1 Receptor 2
- Antigen symbol:IL1R2
- Clonality:Monoclonal
- Clone:8.5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Interleukin 1 Receptor Type II
Clonality: monoclonal
Clone: 8.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human