Anti-IRF4 Rat Monoclonal Antibody
USBII7662-71P
- Antibody type:Primary
- Antigen name:Interferon regulatory factor 4
- Antigen symbol:IRF4
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 10 ?g/ml
Type: Primary
Antigen: IRF4 (interferon regulatory factor 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Human, Mouse