Anti-HLA Class 2 Antigen DR beta Mouse Monoclonal Antibody [clone: 8L293]
Catalog # USBIH6099-96N
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR beta
- Clonality:Monoclonal
- Clone:8L293
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~0,7 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10-1:1000; ELISA: 1:100-1:2000; Western Blot: 1:100-1:2000; Immunochemistry: 1:10-1:500
Type: Primary
Antigen: HLA Class 2 Antigen DR beta
Clonality: monoclonal
Clone: 8L293
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: