Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5F62]
Catalog # USBIH6099-95L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:5F62
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 Tests
- Pk:50 Tests
Specifications
About this item
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 5F62
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human