Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: MEM-136]
USBIH6099-85L
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DP, DR
- Clonality:Monoclonal
- Clone:MEM-136
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 15mM, sodium azide, pH 7,4.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA Class 2 Antigen DP, DR
Clonality: monoclonal
Clone: MEM-136
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human