Anti-HLA Class 2 Antigen DP, DR Mouse Monoclonal Antibody [clone: 6D460]
Catalog # USBIH6098-72B
Leverancier: US Biological
Specificaties
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DP, DR
- Clonality:Monoclonal
- Clone:6D460
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig,Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specificaties
Over dit artikel
Type: Primary
Antigen: HLA Class 2 Antigen DP, DR
Clonality: monoclonal
Clone: 6D460
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Pig