Anti-GFP Mouse monoclonal antibody [clone: 10C303]
Catalog # USBIG8965-22
Supplier: US Biological
Specifications
- Type antilichaam:Label
- antigeen symbool:GFP
- Clonality:Monoklonaal
- Clone:10C303
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Formulier:liquid
- Aminozuurgetal:1 to 238
- Opslagbuffer:PBS, pH 7,2, 0,1% sodium azide.
- Opslagtemperatuur:Cool
- Concentratie:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Maat:100 µl
- VPE:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone: 10C303
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human