Anti-Glycogen Phosphorylase, Muscle Form Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIG8169-96B1FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Glycogen Phosphorylase, Muscle Form
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Glycogen Phosphorylase, Muscle Form
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human